MilliporeSigma
  • Antibody against tuberlin: the specific visualization of cytoplasmic microtubules in tissue culture cells.
Antibody against tuberlin: the specific visualization of cytoplasmic microtubules in tissue culture cells.
Proceedings of the National Academy of Sciences of the United States of America (1975-02-01)
K Weber, R Pollack, T Bibring
PMID804694
ABSTRACT
Cytoplasmic microtubules in tissue culture cells can be directly visualized by immunofluorescence microscopy. Antibody against tubulin from the outer doublets of sea urchin sperm flagella decorates a network of fine cytoplasmic fibers in a variety of cell lines of human, monkey, rat, mouse, and chicken origin. These fibers are separate and of uniform thickness and are seen throughout the cytoplasm. The fibers disappear either in a medium containing colchicine or after subjection of the cells to low temperature. The same treatments do not destroy the microfilamentous structures that are visualized by means of antibody against actin. When tryspin-treated enucleated cells are replated and then stained with antibody against tubulin, the fibers can be seen to traverse the entire enucleated cytoplasm.