MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
3D Bioprinting Using a Templated Porous Bioink.
Advanced healthcare materials (2016-04-30)
James P K Armstrong, Madeline Burke, Benjamin M Carter, Sean A Davis, Adam W Perriman
ABSTRACT
3D tissue printing with adult stem cells is reported. A novel cell-containing multicomponent bioink is used in a two-step 3D printing process to engineer bone and cartilage architectures.