MilliporeSigma
  • Acute and chronic effects of cholinesterase inhibitors and pilocarpine on the density and sensitivity of central and peripheral alpha 2-adrenoceptors.
Acute and chronic effects of cholinesterase inhibitors and pilocarpine on the density and sensitivity of central and peripheral alpha 2-adrenoceptors.
European journal of pharmacology (1993-06-04)
G Olmos, A Miralles, J A García-Sevilla, M T Giralt, J Pineda, M Asier Garro, L Ugedo, A Menargues, R Obach
ABSTRACT
The specific binding of the agonists [3H]clonidine and [3H]UK 14304 (bromoxidine) and of the antagonist [3H]RX 821002 (2-metoxy idazoxan) to rat brain membranes, as well as clonidine-induced mydriasis, clonidine-induced inhibition of brain (3,4-dihydroxyphenylalaninme) DOPA synthesis and clonidine-induced inhibition of twitch responses in the vas deferens, was used to evaluate the density and sensitivity of central and peripheral alpha 2-adrenoceptors after prolonged activation of the cholinergic system. Acute (12 h), short-term (4 days) or chronic (7-18 days) treatment with the cholinesterase inhibitors neostigmine (0.1 mg/kg), physostigmine (0.1 mg/kg) and diisopropylfluorophosphate (2 mg/kg) and with the muscarinic receptor agonist pilocarpine (10 mg/kg) did not alter the density of brain alpha 2-adrenoceptors. In contrast, various functional responses mediated by central and peripheral alpha 2-adrenoceptors were potentiated after the repeated treatments. Thus, the inhibitory alpha 2-autoreceptor that modulates the synthesis of brain noradrenaline and the central postsynaptic inhibitory alpha 2-adrenoceptor that induces mydriasis displayed greater responses in vivo after chronic treatment with neostigmine or pilocarpine. These treatments also increased the sensitivity of peripheral presynaptic alpha 2-adrenoceptors in the vas deferens. The results indicate that prolonged activation of central and peripheral cholinergic pathways results in up-regulation of alpha 2-adrenoceptor function without apparent increases in receptor density.