MilliporeSigma
  • Candida antarctica Lipase-Catalyzed Doubly Enantioselective Aminolysis Reactions. Chemoenzymatic Synthesis of 3-Hydroxypyrrolidines and 4-(Silyloxy)-2-oxopyrrolidines with Two Stereogenic Centers.
Candida antarctica Lipase-Catalyzed Doubly Enantioselective Aminolysis Reactions. Chemoenzymatic Synthesis of 3-Hydroxypyrrolidines and 4-(Silyloxy)-2-oxopyrrolidines with Two Stereogenic Centers.
The Journal of organic chemistry (2001-10-25)
Víctor M. Sánchez, Francisca Rebolledo, Vicente Gotor
ABSTRACT
Aminolyses of racemic and prochiral 3-hydroxyesters with racemic amines are effectively catalyzed by Candida antarctica lipase. In these processes, the simultaneous resolution of the ester and the amine, or the corresponding asymmetrization-resolution, takes place. In all cases, the high enantioselectivity shown by the lipase toward all the reactants allows the preparation of enantiopure 3-hydroxyamides with very high diastereomeric ratios. These 3-hydroxyamides are used as starting materials in the synthesis of 3-hydroxy- and 2-oxopyrrolidines, both containing two stereogenic centers in their structures.