MilliporeSigma
At the crossroads of chemistry and immunology: catalytic antibodies.
Science (New York, N.Y.) (1991-05-03)
R A Lerner, S J Benkovic, P G Schultz
ABSTRACT
Immunochemistry has historically focused on the nature of antigenicity and antibody-antigen recognition. However, in the last 5 years, the field of immunochemistry has taken a new direction. With the aid of mechanistic and synthetic chemistry, the vast network of molecules and cells of the immune system has been tapped to produce antibodies with a new function--catalytic antibodies. Because antibodies can be generated that selectively bind almost any molecule of interest, this new technology offers the potential to tailor-make highly selective catalysts for applications in biology, chemistry, and medicine. In addition, catalytic antibodies provide fundamental insight into important aspects of biological catalysis, including the importance of transition-state stabilization, proximity effects, general acid and base catalysts, electrophilic and nucleophilic catalysis, and strain.