MilliporeSigma
  • Efficient asymmetric epoxidation of alpha,beta-unsaturated ketones using a soluble triblock polyethylene glycol-polyamino acid catalyst.
Efficient asymmetric epoxidation of alpha,beta-unsaturated ketones using a soluble triblock polyethylene glycol-polyamino acid catalyst.
Organic letters (2001-03-22)
R W Flood, T P Geller, S A Petty, S M Roberts, J Skidmore, M Volk
ABSTRACT
[reaction: see text]. Polyethylene glycol (PEG)-bound poly-L-leucine acts as a THF-soluble catalyst for the Juliá-Colonna asymmetric epoxidation of enones. Excellent enantioselectivities may be obtained even with short chain length polyleucine. FT-IR investigations have determined that the catalytically active polyleucine components of these copolymers have an alpha-helical structure.