MilliporeSigma
  • Neuronal expression of caspase-1 immunoreactivity in the rat central nervous system.
Neuronal expression of caspase-1 immunoreactivity in the rat central nervous system.
Journal of neuroimmunology (2003-12-31)
Catharina Lindberg, Charlotta Eriksson, Anne-Marie Van Dam, Bengt Winblad, Marianne Schultzberg
ABSTRACT
Caspase-1/interleukin-1beta (IL-1beta)-converting enzyme (ICE) cleaves IL-1beta and IL-18 precursor proteins to the active forms of these proinflammatory cytokines. Since both cytokines are constitutively expressed in the brain, we investigated whether this is also the case for caspase-1. Using an antibody raised against the p10-subunit of the active enzyme, constitutive expression of caspase-1 immunoreactivity was found in nerve cells in the arcuate nucleus and in nerve fibres throughout the brain. Co-localisation with alpha-melanocyte stimulating hormone was demonstrated. The distribution pattern of caspase-1 immunoreactive structures is consistent with a role to produce mature IL-1beta in regions where IL-1beta mediates fever and sleep.