MilliporeSigma
BLNK required for coupling Syk to PLC gamma 2 and Rac1-JNK in B cells.
Immunity (1999-02-19)
M Ishiai, M Kurosaki, R Pappu, K Okawa, I Ronko, C Fu, M Shibata, A Iwamatsu, A C Chan, T Kurosaki
ABSTRACT
Signaling through the B cell receptor (BCR) is essential for B cell function and development. Despite the key role of Syk in BCR signaling, little is known about the mechanism by which Syk transmits downstream effectors. BLNK (B cell LiNKer protein), a substrate for Syk, is now shown to be essential in activating phospholipase C (PLC)gamma 2 and JNK. The BCR-induced PLC gamma 2 activation, but not the JNK activation, was restored by introduction of PLC gamma 2 membrane-associated form into BLNK-deficient B cells. As JNK activation requires both Rac1 and PLC gamma 2, our results suggest that BLNK regulates the Rac1-JNK pathway, in addition to modulating PLC gamma 2 localization.