MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Delivery of Flavonoids and Saponins from Black Bean (Phaseolus vulgaris) Seed Coats Incorporated into Whole Wheat Bread.
Delivery of Flavonoids and Saponins from Black Bean (Phaseolus vulgaris) Seed Coats Incorporated into Whole Wheat Bread.
International journal of molecular sciences (2016-02-24)
Rocio A Chávez-Santoscoy, Marco A Lazo-Vélez, Sergio O Serna-Sáldivar, Janet A Gutiérrez-Uribe
ABSTRACT
Cereal-based products can be used as vehicles for the delivery of relevant bioactive compounds since they are staple foods for most cultures throughout the world. The health promoting benefits of flavonoids and saponins contained in black bean seed coats have been previously described. In the present work, the effect of adding flavonoids and saponins from black bean seed coat to the typical yeast-leavened whole wheat bread formulation in terms of bread features, organoleptic properties and phytochemical profile was studied. The retention of bioactive compounds was determined and the inhibitory effects of in vitro enzyme digested samples on two colon cancer cell lines (Caco-2 and HT29) was evaluated. The addition of bioactive compounds did not significantly affect baking properties or texture parameters. Among organoleptic properties of enriched breads, only crumb color was affected by the addition of bioactive compounds. However, the use of whole wheat flour partially masked the effect on color. More than 90% of added flavonoids and saponins and 80% of anthocyanins were retained in bread after baking. However, saponins were reduced more than 50% after the in vitro enzyme digestion. The black bean seed coat phytochemicals recovered after in vitro enzyme digestion of enriched breads significantly reduced by 20% the viability of colon cancer cells without affecting standard fibroblast cells (p < 0.05).
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Soyasaponin I