MilliporeSigma
Catalytic asymmetric bromination and chlorination of beta-ketoesters.
Chemistry (Weinheim an der Bergstrasse, Germany) (2004-04-28)
Mauro Marigo, Nagaswami Kumaragurubaran, Karl Anker Jørgensen
ABSTRACT
The first general catalytic asymmetric bromination and chlorination of beta-ketoesters has been developed. The reactions proceed for both acyclic and cyclic beta-ketoesters catalyzed by chiral bisoxazolinecopper(II) complexes giving the corresponding optically active alpha-bromo- and alpha-chloro-beta-ketoesters in high yields and moderate to good enantioselectivities. For the optically active chlorinated products the isolated yields are in the range of 88-99 % and the enantiomeric excesses up to 77 % ee, while the optically active brominated adducts are formed in 70-99 % isolated yield and up to 82 % ee. Based on the absolute configuration of the optically active products, the face selectivity for the catalytic enantioselective halogenation is discussed based on a bidentate coordination of the beta-ketoester to the chiral catalyst and a X-ray structure of chiral alpha,gamma-diketoesterenolatebisoxazolinecopper(II) complex.
Sigma-Aldrich
2,2′-Isopropylidenebis[(4S)-4-tert-butyl-2-oxazoline], 99%
Pricing and availability is not currently available.