MilliporeSigma
  • Parathyroid hormone mediates hematopoietic cell expansion through interleukin-6.
Parathyroid hormone mediates hematopoietic cell expansion through interleukin-6.
PloS one (2010-11-05)
Flavia Q Pirih, Megan N Michalski, Sun W Cho, Amy J Koh, Janice E Berry, Eduardo Ghaname, Pachiyappan Kamarajan, Edith Bonnelye, Charles W Ross, Yvonne L Kapila, Pierre Jurdic, Laurie K McCauley
ABSTRACT
Parathyroid hormone (PTH) stimulates hematopoietic cells through mechanisms of action that remain elusive. Interleukin-6 (IL-6) is upregulated by PTH and stimulates hematopoiesis. The purpose of this investigation was to identify actions of PTH and IL-6 in hematopoietic cell expansion. Bone marrow cultures from C57B6 mice were treated with fms-like tyrosine kinase-3 ligand (Flt-3L), PTH, Flt-3L plus PTH, or vehicle control. Flt-3L alone increased adherent and non-adherent cells. PTH did not directly impact hematopoietic or osteoclastic cells but acted in concert with Flt-3L to further increase cell numbers. Flt-3L alone stimulated proliferation, while PTH combined with Flt-3L decreased apoptosis. Flt-3L increased blasts early in culture, and later increased CD45(+) and CD11b(+) cells. In parallel experiments, IL-6 acted additively with Flt-3L to increase cell numbers and IL-6-deficient bone marrow cultures (compared to wildtype controls) but failed to amplify in response to Flt-3L and PTH, suggesting that IL-6 mediated the PTH effect. In vivo, PTH increased Lin(-) Sca-1(+)c-Kit(+) (LSK) hematopoietic progenitor cells after PTH treatment in wildtype mice, but failed to increase LSKs in IL-6-deficient mice. In conclusion, PTH acts with Flt-3L to maintain hematopoietic cells by limiting apoptosis. IL-6 is a critical mediator of bone marrow cell expansion and is responsible for PTH actions in hematopoietic cell expansion.
Sigma-Aldrich
Tetrahydrofurfuryl acrylate, contains hydroquinone as inhibitor, monomethyl ether hydroquinone as inhibitor