MilliporeSigma
Textile dye dermatitis. A review.
Journal of the American Academy of Dermatology (1985-06-01)
K L Hatch, H I Maibach
ABSTRACT
The occurrence of dermatologic problems caused by consumer exposure to dyes on clothing is reviewed. Thirty-one dyes, mainly disperse with anthraquinone or azo structures, have caused allergic contact dermatitis. Phototoxic dye dermatitis is rare.
Product Description
Supelco
Disperse Blue 124, suitable for microscopy