MilliporeSigma
  • Modified nucleosides: an accurate tumour marker for clinical diagnosis of cancer, early detection and therapy control.
Modified nucleosides: an accurate tumour marker for clinical diagnosis of cancer, early detection and therapy control.
British journal of cancer (2006-05-11)
A Seidel, S Brunner, P Seidel, G I Fritz, O Herbarth
ABSTRACT
Modified nucleosides, regarded as indicators for the whole-body turnover of RNAs, are excreted in abnormal amounts in the urine of patients with malignancies. To test their usefulness as tumour markers and to compare them with the conventional tumour markers, fractionated urine samples were analysed using chromatography. The excretion patterns of nucleosides of 68 cancer patients with malignant and benign tumours and 41 healthy controls have been studied. Significant elevations in the total sum and the concentrations of at least three (or four) of indicator nucleosides cytidine, pseudouridine, 2-pyridone-5-carboxamide-N1-ribofuranoside, N2,N2-dimethylguanine, 1-methylguanosine, 2-methylguanosine and 1-methyladenosine indicate a tumour with a sensitivity of 54% (77%) and a specificity of 86% (98%). Using an artificial neural network analysis, a sensitivity of 97% and a specificity of 85% were achieved in differentiating between tumour and control volunteers. The comparison with carcinoembryonic antigen, cancer antigen 15-3 und tissue polypeptide antigen indicates that urinary nucleosides may be useful tumour markers. This study suggests that the simultaneous determination of modified nucleosides and creatinine in urine samples of patients with cancer leads to an advantage to current methods and is a useful method to detect cancer early and to control the success of therapy.
Sigma-Aldrich
Octadecylsilane, 97%
Sigma-Aldrich
Xanthosine