MilliporeSigma
  • Study on cytochrome p-450 dependent retinoic Acid metabolism and its inhibitors as potential agents for cancer therapy.
Study on cytochrome p-450 dependent retinoic Acid metabolism and its inhibitors as potential agents for cancer therapy.
Scientia pharmaceutica (2011-12-07)
Mobasher Ahmad
ABSTRACT
The relative lack of clinical success with conventional anticancer agents may be due in part to the traditional concept of cancer being a biological state rather than a dynamic process. Redefining cancer as a dynamic disease commencing with carcinogenesis introduces the possibility of chemoprevention. Retinoids offer the promise of a therapeutic option based on differentiation of premalignant as well as malignant cells. Research to date has concentrated on the use of exogenous retinoids in cancer. Although this research continues with new retinoid derivatives, an alternative approach to overcoming the drawbacks associated with exogenous retinoids has been to increase the levels of endogenous retinoic acid (RA) by inhibiting the cytochrome P450- mediated catabolism of RA using a novel class of agents known as retinoic acid metabolism blocking agents (RAMBAs which increase the level of endogenous retinoic acid (RA) within the tumor cells by blocking their metabolism. This approach presents several theoretic advantages.In the present study a wide range of established P-450 inhibitors has been screened to examine their inhibitory activity on all-trans-Retinoic acid (ATRA) metabolism. Forty-one known P450 inhibitors were tested for their inhibitory activity against RA metabolism. Most of them are nitrogen-containing compounds. The results showed that among these compounds only six compounds (N-benzyl-2-phenylethanamine, itraconazole, chlorpromazine, 5-chloro-1,3-benzoxazol-2-amine, proadifen and furazolidone) showed inhibition of RA metabolism which was > 50%. Ketoconazole and liarozole were also screened as standard potent inhibitors in the same system and gave 87.5% and 89% inhibition, respectively. The results indicate that mostly azoles with substituents in positions other than the 1-position on the ring are very weak inhibitors of RA metabolism. The most effective inhibitors (ketoconazole, itraconazole, bifonazole and clotrimazole) are 1-substituted and possess relatively large aromatic groups in the molecule. 1-Substituted imidazoles bind to cytochrome P-450 with a very high affinity but substitution in the other position of the imidazole decreases the binding affinity.