MilliporeSigma
  • (D)-Amino acid chemical reporters reveal peptidoglycan dynamics of an intracellular pathogen.
(D)-Amino acid chemical reporters reveal peptidoglycan dynamics of an intracellular pathogen.
ACS chemical biology (2012-12-18)
M Sloan Siegrist, Sarah Whiteside, John C Jewett, Arjun Aditham, Felipe Cava, Carolyn R Bertozzi
ABSTRACT
Peptidoglycan (PG) is an essential component of the bacterial cell wall. Although experiments with organisms in vitro have yielded a wealth of information on PG synthesis and maturation, it is unclear how these studies translate to bacteria replicating within host cells. We report a chemical approach for probing PG in vivo via metabolic labeling and bioorthogonal chemistry. A wide variety of bacterial species incorporated azide and alkyne-functionalized d-alanine into their cell walls, which we visualized by covalent reaction with click chemistry probes. The d-alanine analogues were specifically incorporated into nascent PG of the intracellular pathogen Listeria monocytogenes both in vitro and during macrophage infection. Metabolic incorporation of d-alanine derivatives and click chemistry detection constitute a facile, modular platform that facilitates unprecedented spatial and temporal resolution of PG dynamics in vivo.
Sigma-Aldrich
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97%