MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • The effect of primary organic particles on emergency hospital admissions among the elderly in 3 US cities.
The effect of primary organic particles on emergency hospital admissions among the elderly in 3 US cities.
Environmental health : a global access science source (2013-08-29)
Marianthi-Anna Kioumourtzoglou, Antonella Zanobetti, Joel D Schwartz, Brent A Coull, Francesca Dominici, Helen H Suh
ABSTRACT
Fine particle (PM2.5) pollution related to combustion sources has been linked to a variety of adverse health outcomes. Although poorly understood, it is possible that organic carbon (OC) species, particularly those from combustion-related sources, may be partially responsible for the observed toxicity of PM2.5. The toxicity of the OC species may be related to their chemical structures; however, few studies have examined the association of OC species with health impacts. We categorized 58 primary organic compounds by their chemical properties into 5 groups: n-alkanes, hopanes, cyclohexanes, PAHs and isoalkanes. We examined their impacts on the rate of daily emergency hospital admissions among Medicare recipients in Atlanta, GA and Birmingham, AL (2006-2009), and Dallas, TX (2006-2007). We analyzed data in two stages; we applied a case-crossover analysis to simultaneously estimate effects of individual OC species on cause-specific hospital admissions. In the second stage we estimated the OC chemical group-specific effects, using a multivariate weighted regression. Exposures to cyclohexanes of six days and longer were significantly and consistently associated with increased rate of hospital admissions for CVD (3.40%, 95%CI = (0.64, 6.24%) for 7-d exposure). Similar increases were found for hospitalizations for ischemic heart disease and myocardial infarction. For respiratory related hospital admissions, associations with OC groups were less consistent, although exposure to iso-/anteiso-alkanes was associated with increased respiratory-related hospitalizations. Results suggest that week-long exposures to traffic-related, primary organic species are associated with increased rate of total and cause-specific CVD emergency hospital admissions. Associations were significant for cyclohexanes, but not hopanes, suggesting that chemical properties likely play an important role in primary OC toxicity.
Product Description
Supelco
17β(H),21β(H)-Hopane solution, 0.1 mg/mL in isooctane, analytical standard