MilliporeSigma
  • Apoptosis signal-regulating kinase 1 is involved in WISP-1-promoted cell motility in human oral squamous cell carcinoma cells.
Apoptosis signal-regulating kinase 1 is involved in WISP-1-promoted cell motility in human oral squamous cell carcinoma cells.
PloS one (2013-11-10)
Jing-Yuan Chuang, An-Chen Chang, I-Ping Chiang, Ming-Hsui Tsai, Chih-Hsin Tang
ABSTRACT
Oral squamous cell carcinoma (OSCC) has a tendency to migrate and metastasize. WNT1-inducible signaling pathway protein 1 (WISP-1) is a cysteine-rich protein that belongs to the Cyr61, CTGF, Nov (CCN) family of matrix cellular proteins. The effect of WISP-1 on human OSCC cells, however, is unknown. Here, we showed that WISP-1 increased cell migration and intercellular adhesion molecule-1 (ICAM-1) expression in OSCC cells. Pretreatment of cells with integrin αvβ3 monoclonal antibody (mAb) significantly abolished WISP-1-induced cell migration and ICAM-1 expression. On the other hand, WISP-1-mediated cell motility and ICAM-1 upregulation were attenuated by ASK1, JNK, and p38 inhibitor. Furthermore, WISP-1 also enhanced activator protein 1 (AP-1) activation, and the integrin αvβ3 mAb, and ASK1, JNK, and p38 inhibitors reduced WISP-1-mediated AP-1 activation. Moreover, WISP-1 and ICAM-1 expression correlated with the tumor stage of patients with OSCC. Our results indicate that WISP-1 enhances the migration of OSCC cells by increasing ICAM-1 expression through the αvβ3 integrin receptor and the ASK1, JNK/p38, and AP-1 signal transduction pathways.
Sigma-Aldrich
MISSION® esiRNA, targeting human WISP1