MilliporeSigma
  • Newly discovered tachykinins raise new questions about their peripheral roles and the tachykinin nomenclature.
Newly discovered tachykinins raise new questions about their peripheral roles and the tachykinin nomenclature.
Trends in pharmacological sciences (2004-01-16)
Riccardo Patacchini, Alessandro Lecci, Peter Holzer, Carlo Alberto Maggi
ABSTRACT
The tachykinin family has recently been extended by the discovery of a third tachykinin gene encoding previously unknown mammalian tachykinins (hemokinin 1, endokinin A and endokinin B) that have a widespread peripheral distribution and a tachykinin NK(1) receptor selectivity. This and the identification of other tachykinin-like peptides such as C14TKL-1 and virokinin raise many questions about the roles played by tachykinins in peripheral tissues and render terms such as 'neurokinins' and 'SP receptor' inappropriate.