MilliporeSigma
  • Senataxin, defective in ataxia oculomotor apraxia type 2, is involved in the defense against oxidative DNA damage.
Senataxin, defective in ataxia oculomotor apraxia type 2, is involved in the defense against oxidative DNA damage.
The Journal of cell biology (2007-06-15)
Amila Suraweera, Olivier J Becherel, Philip Chen, Natalie Rundle, Rick Woods, Jun Nakamura, Magtouf Gatei, Chiara Criscuolo, Alessandro Filla, Luciana Chessa, Markus Fusser, Bernd Epe, Nuri Gueven, Martin F Lavin
ABSTRACT
A defective response to DNA damage is observed in several human autosomal recessive ataxias with oculomotor apraxia, including ataxia-telangiectasia. We report that senataxin, defective in ataxia oculomotor apraxia (AOA) type 2, is a nuclear protein involved in the DNA damage response. AOA2 cells are sensitive to H2O2, camptothecin, and mitomycin C, but not to ionizing radiation, and sensitivity was rescued with full-length SETX cDNA. AOA2 cells exhibited constitutive oxidative DNA damage and enhanced chromosomal instability in response to H2O2. Rejoining of H2O2-induced DNA double-strand breaks (DSBs) was significantly reduced in AOA2 cells compared to controls, and there was no evidence for a defect in DNA single-strand break repair. This defect in DSB repair was corrected by full-length SETX cDNA. These results provide evidence that an additional member of the autosomal recessive AOA is also characterized by a defective response to DNA damage, which may contribute to the neurodegeneration seen in this syndrome.
Sigma-Aldrich
Diethyl 3,4-pyrroledicarboxylate, 98%