MilliporeSigma
  • The enantiomers of tramadol and its major metabolite inhibit peristalsis in the guinea pig small intestine via differential mechanisms.
The enantiomers of tramadol and its major metabolite inhibit peristalsis in the guinea pig small intestine via differential mechanisms.
BMC pharmacology (2007-03-21)
Michael K Herbert, Rebecca Weis, Peter Holzer
ABSTRACT
Inhibition of intestinal peristalsis is a major side effect of opioid analgesics. Although tramadol is an opioid-like analgesic, its effect on gut motility is little known. Therefore, the effect of (+)-tramadol, (-)-tramadol and the major metabolite O-desmethyltramadol on intestinal peristalsis in vitro and their mechanisms of action were examined. Distension-induced peristalsis was recorded in fluid-perfused segments of the guinea pig small intestine. The intraluminal peristaltic pressure threshold (PPT) was used to quantify the motor effects of extraserosally administered drugs. Racemic tramadol, its (+)- and (-)-enantiomers and the major metabolite O-desmethyltramadol (0.1-100 microM) concentration-dependently increased PPT until peristalsis was transiently or persistently abolished. The rank order of potency was (-)-tramadol < (+)-tramadol <O-desmethyltramadol. The peristaltic motor inhibition caused by (+)- and (-)-tramadol was markedly and that of O-desmethyltramadol nearly completely prevented by naloxone, but left unaltered by the 5-hydroxytryptamine receptor antagonists methysergide plus tropisetron. The adrenoceptor antagonists prazosin plus yohimbine reduced the effect of (+)- and (-)-tramadol but not that of O-desmethyltramadol. The results show that the metabolite O-desmethyltramadol is more potent in inhibiting peristalsis than its parent compound. The action of all tramadol forms depends on opioid receptors, and that of (+)- and (-)-tramadol also involves adrenoceptors.