MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • T cells can use either T cell receptor or CD28 receptors to absorb and internalize cell surface molecules derived from antigen-presenting cells.
T cells can use either T cell receptor or CD28 receptors to absorb and internalize cell surface molecules derived from antigen-presenting cells.
The Journal of experimental medicine (2000-04-05)
I Hwang, J F Huang, H Kishimoto, A Brunmark, P A Peterson, M R Jackson, C D Surh, Z Cai, J Sprent
ABSTRACT
At the site of contact between T cells and antigen-presenting cells (APCs), T cell receptor (TCR)-peptide-major histocompatibility complex (MHC) interaction is intensified by interactions between other molecules, notably by CD28 and lymphocyte function-associated antigen 1 (LFA-1) on T cells interacting with B7 (B7-1 and B7-2), and intracellular adhesion molecule 1 (ICAM-1), respectively, on APCs. Here, we show that during T cell-APC interaction, T cells rapidly absorb various molecules from APCs onto the cell membrane and then internalize these molecules. This process is dictated by at least two receptors on T cells, namely CD28 and TCR molecules. The biological significance of T cell uptake of molecules from APCs is unclear. One possibility is that this process may allow activated T cells to move freely from one APC to another and eventually gain entry into the circulation.
Sigma-Aldrich
trans-2-Bromovinylboronic acid MIDA ester, technical grade