MilliporeSigma
  • Efficient activation of reconstructed rat embryos by cyclin-dependent kinase inhibitors.
Efficient activation of reconstructed rat embryos by cyclin-dependent kinase inhibitors.
PloS one (2010-03-25)
Robin L Webb, Kirk A Findlay, Michael A Green, Tina L Beckett, M Paul Murphy
ABSTRACT
Over the last decade a number of species, from farm animals to rodents, have been cloned using somatic cell nuclear transfer technology (SCNT). This technique has the potential to revolutionize the way that genetically modified animals are made. In its current state, the process of SCNT is very inefficient (<5% success rate), with several technical and biological hurdles hindering development. Yet, SCNT provides investigators with powerful advantages over other approaches, such as allowing for prescreening for the desired level of transgene expression and eliminating the excess production of undesirable wild-type animals. The rat plays a significant role in biomedical research, but SCNT has been problematic for this species. In this study, we address one aspect of the problem by evaluating methods of activation in artificially constructed rat embryos. We demonstrate that treatment with a calcium ionophore (ionomycin) combined with a variety of cyclin-dependent kinase inhibitors is an effective way to activate rat embryos. This is in contrast to methods developed for the mouse embryo, which tolerates much less specific chemical treatments. Methods developed to activate mouse embryos do not translate well to rat embryos. Activation methods developed for one species will not necessarily translate to another species, even if it is closely related. Further, the parthenogenic response to chemical activators is not always a reliable indicator of how reconstructed embryos will react to the same activation method. A better understanding of rat oocyte physiology, although essential for developing better models of disease, may also provide insights that will be useful for making the SCNT process more efficient.
Sigma-Aldrich
6-(Dimethylamino)purine, ≥98%