MilliporeSigma
  • A general, convergent strategy for the construction of indolizidine alkaloids: total syntheses of (-)-indolizidine 223AB and alkaloid (-)-205B.
A general, convergent strategy for the construction of indolizidine alkaloids: total syntheses of (-)-indolizidine 223AB and alkaloid (-)-205B.
The Journal of organic chemistry (2006-03-25)
Amos B Smith, Dae-Shik Kim
ABSTRACT
N-Toluenesulfonyl aziridines comprise effective second electrophiles in the solvent controlled three-component linchpin union of silyl dithianes for the stereocontrolled convergent elaboration of protected 1,5-amino alcohols. This tactic, in conjunction with a one-flask sequential cyclization, constitutes an effective general strategy for the construction of indolizidine and related alkaloids, illustrated here with the total syntheses of (-)-indolizidine 223AB (1) and alkaloid (-)-205B (2).