MilliporeSigma
  • Asymmetric hydrogenation of itaconic acid and enol acetate derivatives with the Rh-TangPhos catalyst.
Asymmetric hydrogenation of itaconic acid and enol acetate derivatives with the Rh-TangPhos catalyst.
Organic letters (2003-01-17)
Wenjun Tang, Duan Liu, Xumu Zhang
ABSTRACT
[reaction: see text] The Rh-TangPhos catalyst has been used for asymmetric hydrogenation of itaconic acid and enol acetate derivatives. A variety of chiral 2-substituted succinic acids and chiral acetates have been obtained in excellent ee values (up to 99% ee).