MilliporeSigma
  • A bisphosphepine ligand with stereogenic phosphorus centers for the practical synthesis of beta-aryl-beta-amino acids by asymmetric hydrogenation.
A bisphosphepine ligand with stereogenic phosphorus centers for the practical synthesis of beta-aryl-beta-amino acids by asymmetric hydrogenation.
Angewandte Chemie (International ed. in English) (2003-08-06)
Wenjun Tang, Weimin Wang, Yongxiang Chi, Xumu Zhang