MilliporeSigma
  • Transition metal-catalyzed intermolecular [5+2] and [5+2+1] cycloadditions of allenes and vinylcyclopropanes.
Transition metal-catalyzed intermolecular [5+2] and [5+2+1] cycloadditions of allenes and vinylcyclopropanes.
Journal of the American Chemical Society (2005-05-05)
Hermann A Wegner, Armin de Meijere, Paul A Wender
ABSTRACT
Initial examples of the intermolecular Rh(I)-catalyzed [5+2] cycloaddition reaction of bifunctional allenes and vinylcyclopropanes are described. The reactions proceed with facility and in yields of up to 99% with a variety of alkyne-, ester-, styrene-, or cyano-substituents on the allene to afford the corresponding cycloadducts. In the presence of CO, the reaction proceeds to an eight-membered ring cycloadduct and its transannularly closed product, providing the first example of a three-component [5+2+1] cycloaddition with allenes.