MilliporeSigma
  • An antibody to de-N-acetyl sialic acid containing-polysialic acid identifies an intracellular antigen and induces apoptosis in human cancer cell lines.
An antibody to de-N-acetyl sialic acid containing-polysialic acid identifies an intracellular antigen and induces apoptosis in human cancer cell lines.
PloS one (2011-11-19)
Lindsay M Steirer, Gregory R Moe
ABSTRACT
Polysialic acid (PSA), an α2,8-linked homopolymer of N-acetylneuraminic acid (Neu5Ac), is developmentally regulated and its expression is thought to be restricted to a few tissues in adults. Recently, we showed that two human pathogens expressed a derivative of PSA containing de-N-acetyl sialic acid residues (NeuPSA). Here we show that an epitope identified by the anti-NeuPSA monoclonal antibody, SEAM 3 (SEAM 3-reactive antigen or S3RA), is expressed in human melanomas, and also intracellularly in a human melanoma cell line (SK-MEL-28), a human T cell leukemia cell line (Jurkat), and two neuroblastoma cell lines (CHP-134 and SH-SY5Y). SEAM 3 binding induced apoptosis in the four cell lines tested. The unusual intracellular distribution of S3RA was similar to that described for the PSA polysialyltransferases, STX and PST, which are also expressed in the four cell lines used here. Interestingly, suppression of PST mRNA expression by transfection of SK-MEL-28 cells with PST-specific short interfering RNA (siRNA) resulted in decreased SEAM 3 binding. The results suggest further studies of the utility of antibodies such as SEAM 3 as therapeutic agents for certain malignancies.