MilliporeSigma
  • Rhodium nanoparticles entrapped in boehmite nanofibers: recyclable catalyst for arene hydrogenation under mild conditions.
Rhodium nanoparticles entrapped in boehmite nanofibers: recyclable catalyst for arene hydrogenation under mild conditions.
Chemical communications (Cambridge, England) (2005-11-18)
In Soo Park, Min Serk Kwon, Namdu Kim, Jae Sung Lee, Kyung Yeon Kang, Jaiwook Park
ABSTRACT
A new recyclable rhodium catalyst was synthesized by a simple procedure from readily available reagents, which showed high activities in the hydrogenation of various arenes under 1 atm H2 at room temperature.