MilliporeSigma
  • G protein-coupled receptor kinase 2 (GRK2): mechanisms of regulation and physiological functions.
G protein-coupled receptor kinase 2 (GRK2): mechanisms of regulation and physiological functions.
FEBS letters (1998-07-25)
A M Aragay, A Ruiz-Gómez, P Penela, S Sarnago, A Elorza, M C Jiménez-Sainz, F Mayor
ABSTRACT
G protein-coupled receptor kinase 2 (GRK2) plays a key role in determining the rate and extent of G protein-coupled receptor (GPCR) desensitization and resensitization. Recent data indicate that GRK2 activity, subcellular distribution and expression are tightly regulated. The important physiological function of GRK2 as a modulator of the efficacy of GPCR signal transduction systems is exemplified by its relevance in cardiovascular physiopathology as well as by its emerging role in the regulation of chemokine receptors.