MilliporeSigma
  • A catalyst for efficient and highly enantioselective hydrogenation of aromatic, heteroaromatic, and alpha,beta-unsaturated ketones
A catalyst for efficient and highly enantioselective hydrogenation of aromatic, heteroaromatic, and alpha,beta-unsaturated ketones
Organic letters (2001-01-11)
Burk, Hems, Herzberg, Malan, Zanotti-Gerosa
ABSTRACT
PhanePhos-ruthenium-diamine complexes catalyze the asymmetric hydrogenation of a wide range of aromatic, heteroaromatic, and alpha, beta-unsaturated ketones with high activity and excellent enantioselectivity.
Sigma-Aldrich
(R)-(–)-4,12-Bis(diphenylphosphino)-[2.2]-paracyclophane, 96%
Sigma-Aldrich
(S)-(+)-4,12-Bis(diphenylphosphino)-[2.2]-paracyclophane, 96%