MilliporeSigma
  • Suzuki-type cross-coupling reaction of pentavalent triarylantimony diacetates with arylboronic acids without a base
Suzuki-type cross-coupling reaction of pentavalent triarylantimony diacetates with arylboronic acids without a base
Tetrahedron Letters (2007)
Shuji Yasuike, et al.