MilliporeSigma
  • Hydroxytyrosol in functional hydroxytyrosol-enriched biscuits is highly bioavailable and decreases oxidised low density lipoprotein levels in humans.
Hydroxytyrosol in functional hydroxytyrosol-enriched biscuits is highly bioavailable and decreases oxidised low density lipoprotein levels in humans.
Food chemistry (2016-03-24)
Raquel Mateos, Sara Martínez-López, Gema Baeza Arévalo, Miryam Amigo-Benavent, Beatriz Sarriá, Laura Bravo-Clemente
ABSTRACT
Hydroxytyrosol (HT) and its derivatives in olive oil protect low-density lipoproteins (LDL) against oxidation. Biscuits could be a convenient alternative to broaden consumers' choice of HT-rich foods, although the biscuit matrix could affect HT bioavailability. We performed a crossover, randomized, double-blind study to evaluate HT bioavailability in HT-enriched biscuits (HT-B) versus non-enriched biscuits (C-B), and the effects on oxidative postprandial status. On two separate days, 13 subjects consumed 30 g of C-B or HT-B (5.25mg HT) after overnight-fasting. Blood and urine were collected at different intervals and analysed by LC-MS-QToF. After HT-B consumption, plasma metabolites peaked between 0.5 and 1h and were extensively excreted in urine. HT-sulphate and 3,4-dihydroxyphenylacetic acid (DOPAC)-sulphate were the main metabolites, followed by DOPAC and homovanillic acid (HVA). HT-glucuronide, DOPAC-glucuronide, HVA-glucuronide and HVA-sulphate were also detected. Postprandial oxidised-LDL concentrations decreased with HT-B. HT is a promising functional ingredient and, in biscuits, it is highly bioavailable and lowers postprandial oxidised-LDL levels.
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%