- Oral Bacitracin: A Consideration for Suppression of Intestinal Vancomycin-Resistant Enterococci (VRE) and for VRE Bacteremia From an Apparent Gastrointestinal Tract Source.
Oral Bacitracin: A Consideration for Suppression of Intestinal Vancomycin-Resistant Enterococci (VRE) and for VRE Bacteremia From an Apparent Gastrointestinal Tract Source.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America (2015-02-24)
Truc T Tran, Hannah R Palmer, Marion R Weimar, Cesar A Arias, Gregory M Cook, Barbara E Murray
PMID25697741