MilliporeSigma
Genetic inhibition of JNK3 ameliorates spinal muscular atrophy.
Human molecular genetics (2015-10-02)
Naresh K Genabai, Saif Ahmad, Zhanying Zhang, Xiaoting Jiang, Cynthia A Gabaldon, Laxman Gangwani
ABSTRACT
Mutation of the Survival Motor Neuron 1 (SMN1) gene causes spinal muscular atrophy (SMA), an autosomal recessive neurodegenerative disorder that occurs in early childhood. Degeneration of spinal motor neurons caused by SMN deficiency results in progressive muscle atrophy and death in SMA. The molecular mechanism underlying neurodegeneration in SMA is unknown. No treatment is available to prevent neurodegeneration and reduce the burden of illness in SMA. We report that the c-Jun NH2-terminal kinase (JNK) signaling pathway mediates neurodegeneration in SMA. The neuron-specific isoform JNK3 is required for neuron degeneration caused by SMN deficiency. JNK3 deficiency reduces degeneration of cultured neurons caused by low levels of SMN. Genetic inhibition of JNK pathway in vivo by Jnk3 knockout results in amelioration of SMA phenotype. JNK3 deficiency prevents the loss of spinal cord motor neurons, reduces muscle degeneration, improves muscle fiber thickness and muscle growth, improves motor function and overall growth and increases lifespan of mice with SMA that shows a systemic rescue of phenotype by a SMN-independent mechanism. JNK3 represents a potential (non-SMN) therapeutic target for the treatment of SMA.
Sigma-Aldrich
Anti-phospho-MAP3K1 (pThr1402) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-ASK1 (pSer966) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Neurofilament 145 kDa Antibody, CT, clone 3H11, clone 3H11, Chemicon®, from mouse