MilliporeSigma
  • Calcitonin gene-related peptide is a key factor in the homing of transplanted human MSCs to sites of spinal cord injury.
Calcitonin gene-related peptide is a key factor in the homing of transplanted human MSCs to sites of spinal cord injury.
Scientific reports (2016-06-15)
Yu Zhang, Jinhua Yang, Peng Zhang, Tao Liu, Jianwei Xu, Zhihai Fan, Yixin Shen, Wenjie Li, Huanxiang Zhang
ABSTRACT
Mesenchymal stem cells (MSCs) can be used to treat many diseases, including spinal cord injury (SCI). Treatment relies mostly on the precise navigation of cells to the injury site for rebuilding the damaged spinal cord. However, the key factors guiding MSCs to the epicenter of SCI remain unknown. Here, we demonstrated that calcitonin gene-related peptide (CGRP), a neural peptide synthesized in spinal cord, can dramatically aid the homing of human umbilical cord mesenchymal stem cells (HUMSCs) in spinal cord-transected SCI rats. First, HUMSCs exhibited chemotactic responses in vitro to CGRP. By time-lapse video analysis, increased chemotactic index (CMI), forward migration index (FMI) and speed contributed to this observed migration. Then, through enzyme immunoassay, higher CGRP concentrations at the lesion site were observed after injury. The release of CGRP directed HUMSCs to the injury site, which was suppressed by CGRP 8-37, a CGRP antagonist. We also verified that the PI3K/Akt and p38MAPK signaling pathways played a critical role in the CGRP-induced chemotactic migration of HUMSCs. Collectively, our data reveal that CGRP is a key chemokine that helps HUMSCs migrate to the lesion site and thereby can be used as a model molecule to study MSCs homing after SCI.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP)−Cy3 antibody, Mouse monoclonal, clone G-A-5, purified from hybridoma cell culture