MilliporeSigma
  • Arylphthalazines. Part 2: 1-(Isoquinolin-5-yl)-4-arylamino phthalazines as potent inhibitors of VEGF receptors I and II.
Arylphthalazines. Part 2: 1-(Isoquinolin-5-yl)-4-arylamino phthalazines as potent inhibitors of VEGF receptors I and II.
Bioorganic & medicinal chemistry letters (2006-01-03)
Matthew A J Duncton, Evgueni L Piatnitski, Reeti Katoch-Rouse, Leon M Smith, Alexander S Kiselyov, Daniel L Milligan, Chris Balagtas, Wai C Wong, Joel Kawakami, Jacqueline F Doody
ABSTRACT
A novel class of 1-(isoquinolin-5-yl)-4-arylamino-phthalazines is described as inhibitors of vascular endothelial growth factor receptor II (VEGFR-2). Many compounds display VEGFR-2 inhibitory activity with an IC(50) as low as 0.017 microM in an HTRF enzymatic assay. The compounds also inhibit VEGFR-1, a related tyrosine kinase.
Sigma-Aldrich
1,4-Dichlorophthalazine, 95%
Pricing and availability is not currently available.