MilliporeSigma
  • Nitrogen and phosphorus co-doped graphene quantum dots: synthesis from adenosine triphosphate, optical properties, and cellular imaging.
Nitrogen and phosphorus co-doped graphene quantum dots: synthesis from adenosine triphosphate, optical properties, and cellular imaging.
Nanoscale (2015-04-16)
Arundithi Ananthanarayanan, Yue Wang, Parimal Routh, Mahasin Alam Sk, Aung Than, Ming Lin, Jie Zhang, Jie Chen, Handong Sun, Peng Chen
ABSTRACT
Graphene quantum dots (GQDs) are emerging zero-dimensional materials promising a wide spectrum of applications, particularly, as superior fluorescent reporters for bio-imaging and optical sensing. Heteroatom doping can endow GQDs with new or improved photoluminescence properties. Here, we demonstrate a simple strategy for the synthesis of nitrogen and phosphorus co-doped GQDs from a single biomolecule precursor (adenosine triphosphate - ATP). Such ATP-GQDs exhibit high fluorescence quantum yield, strong two-photon upconversion, small molecular weight, high photostability, and good biocompatibility. Furthermore, transferrin conjugated ATP-GQDs have been used for imaging and real-time tracking of transferrin receptors in live cells.
Sigma-Aldrich
Nitrogen/Phosphorus co-doped graphene, avg. no. of layers, 1 ‑ 5
Pricing and availability is not currently available.