MilliporeSigma
  • Synthesis of 9-amino-5, 6, 7, 8-tetrahydrothieno [3, 2-b] quinoline and 9-amino-5, 6, 7, 8-tetrahydrothieno [3, 2-b] quinolin-8-ol derivatives.
Synthesis of 9-amino-5, 6, 7, 8-tetrahydrothieno [3, 2-<I>b</I>] quinoline and 9-amino-5, 6, 7, 8-tetrahydrothieno [3, 2-<I>b</I>] quinolin-8-ol derivatives.
Journal of Heterocyclic Chemistry (2009)
Song YH and Jo BS.