MilliporeSigma
Galanin - a novel biologically active peptide from porcine intestine.
FEBS letters (1983-11-28)
K Tatemoto, A Rökaeus, H Jörnvall, T J McDonald, V Mutt
ABSTRACT
The isolation of a novel biologically active peptide, designated galanin, is described. The peptide was discovered by the detection of its C-terminal amide structure in porcine intestinal extract using a chemical method. It was found that galanin consists of 29 amino acids and the complete amino acid sequence is: Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-His-Ala-Ile-Asp-Asn-His -Arg-Ser -Phe-His-Asp-Lys-Tyr-Gly-Leu-Ala-NH2. Galanin was found to contract smooth muscle preparations from the rat and to cause a mild and sustained hyperglycemia in dog.