MilliporeSigma
Mast cells can enhance resistance to snake and honeybee venoms.
Science (New York, N.Y.) (2006-07-29)
Martin Metz, Adrian M Piliponsky, Ching-Cheng Chen, Verena Lammel, Magnus Abrink, Gunnar Pejler, Mindy Tsai, Stephen J Galli
ABSTRACT
Snake or honeybee envenomation can cause substantial morbidity and mortality, and it has been proposed that the activation of mast cells by snake or insect venoms can contribute to these effects. We show, in contrast, that mast cells can significantly reduce snake-venom-induced pathology in mice, at least in part by releasing carboxypeptidase A and possibly other proteases, which can degrade venom components. Mast cells also significantly reduced the morbidity and mortality induced by honeybee venom. These findings identify a new biological function for mast cells in enhancing resistance to the morbidity and mortality induced by animal venoms.