MilliporeSigma
  • Comparative study of phenol alkylation mechanisms using homogeneous and silica-supported boron trifluoride catalysts.
Comparative study of phenol alkylation mechanisms using homogeneous and silica-supported boron trifluoride catalysts.
J. Mol. Catal. A: Chem. (2000)
Wilson K, et al.