MilliporeSigma
  • Acylation of indoles by Duff reaction and Vilsmeier-Haack formylation and conformation of N-formylindoles.
Acylation of indoles by Duff reaction and Vilsmeier-Haack formylation and conformation of N-formylindoles.
The Journal of Organic Chemistry (1973)
Chatterjee A and Biswas KM.