MilliporeSigma
  • Single nucleotide polymorphisms in the FAM167A-BLK gene are associated with polymyositis/dermatomyositis in the Han Chinese population.
Single nucleotide polymorphisms in the FAM167A-BLK gene are associated with polymyositis/dermatomyositis in the Han Chinese population.
Immunologic research (2015-04-08)
Si Chen, Wei Wu, Jing Li, Qian Wang, Yuan Li, Ziyan Wu, Wenjie Zheng, Qingjun Wu, Chanyuan Wu, Fengchun Zhang, Yongzhe Li
ABSTRACT
Idiopathic inflammatory myopathies (IIMs) are autoimmune diseases influenced by genetic background and environmental factors. Recently, FAM167A-BLK gene has been identified as a potential genetic susceptibility locus for dermatomyositis (DM) in patients of European and Japanese populations. Our aim here was to investigate the association between FAM167A-BLK polymorphisms and IIMs risk in Chinese Han. The FAM167A-BLK single nucleotide polymorphisms (SNPs) rs2736340, rs7812879, rs13277113, rs2618479, rs2254546 and rs2248932 were analyzed in polymyositis (PM) patients (n = 310), DM patients (n = 535) and 968 ethnically matched healthy controls, with the Sequenom MassArray system. Our present study demonstrated that strong allele association was observed in overall PM/DM and PM patients for rs2736340 (P c = 6.48 × 10(-3); P c = 0.013, respectively), rs7812879 (P c = 0.017; P c = 0.034, respectively) and rs13277113 (P c = 0.011; P c = 0.047, respectively). These three SNPs were significantly associated with interstitial lung disease (ILD) in overall PM/DM patients (all, P c < 0.05). The frequency of the five haplotypes of the five SNPs (rs2736340, rs7812879, rs13277113, rs2618479 and rs2254546) was also significantly different between overall PM/DM, PM or DM patients and healthy controls. This was the first study to demonstrate that the FAM167A-BLK polymorphisms were associated with Chinese PM/DM patients or these patients with ILD, indicating that PM/DM might share common gene with other autoimmune diseases.