MilliporeSigma
  • Infrared spectroscopy studies on stability of dimethyl sulfoxide for application in a Li?air battery
Infrared spectroscopy studies on stability of dimethyl sulfoxide for application in a Li?air battery
The Journal of Physical Chemistry C (2013)
Mozhzhukhina N, et al.