MilliporeSigma
A concise total synthesis of melithiazole C.
Organic letters (2007-07-20)
Julian Gebauer, Stellios Arseniyadis, Janine Cossy
ABSTRACT
A short and convergent synthesis of the myxobacterial antibiotic melithiazole C is described featuring a highly E-selective cross-metathesis as the key step.
Sigma-Aldrich
2,4-Dibromothiazole, 97%
Pricing and availability is not currently available.