MilliporeSigma
A nonacid degradable linker for solid-phase synthesis.
Organic letters (2007-09-18)
Aina Colombo, Natalia De la Figuera, Joan Carles Fernàndez, Dolors Fernández-Forner, Fernando Albericio, Pilar Forns
ABSTRACT
Synthesis and applications of two new nonacid degradable linkers as an alternative to the Wang linker for solid-phase synthesis are described. Resin from linker 2 looks superior to linker 1 in terms of yields for both anchoring of the first building block and cleavage and in terms of higher purity of the final product. Use of linker 2 avoids side reactions associated with the use of Wang resin due to an undesired cleavage during final acid treatment.