MilliporeSigma
  • Local administration of platelet-derived growth factor B (PDGFB) improves follicular development and ovarian angiogenesis in a rat model of Polycystic Ovary Syndrome.
Local administration of platelet-derived growth factor B (PDGFB) improves follicular development and ovarian angiogenesis in a rat model of Polycystic Ovary Syndrome.
Molecular and cellular endocrinology (2016-06-04)
Mariana Di Pietro, Leopoldina Scotti, Griselda Irusta, Marta Tesone, Fernanda Parborell, Dalhia Abramovich
ABSTRACT
Alterations in ovarian angiogenesis are common features in Polycystic Ovary Syndrome (PCOS) patients; the most studied of these alterations is the increase in vascular endothelial growth factor (VEGF) production by ovarian cells. Platelet-derived growth factor B (PDGFB) and D (PDGFD) are decreased in follicular fluid of PCOS patients and in the ovaries of a rat model of PCOS. In the present study, we aimed to analyze the effects of local administration of PDGFB on ovarian angiogenesis, follicular development and ovulation in a DHEA-induced PCOS rat model. Ovarian PDGFB administration to PCOS rats partially restored follicular development, decreased the percentage of cysts, increased the percentage of corpora lutea, and decreased the production of anti-Müllerian hormone. In addition, PDGFB administration improved ovarian angiogenesis by reversing the increase in periendothelial cell area and restoring VEGF levels. Our results shed light into the mechanisms that lead to altered ovarian function in PCOS and provide new data for potential therapeutic strategies.
Sigma-Aldrich
Normal Mouse IgG, Alexa Fluor 488 conjugate, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunofluorescence.