MilliporeSigma
  • Modulation of macrophage functionality induced in vitro by chlorpyrifos and carbendazim pesticides.
Modulation of macrophage functionality induced in vitro by chlorpyrifos and carbendazim pesticides.
Journal of immunotoxicology (2016-07-19)
Imen Helali, Saiida Ferchichi, Amal Maaouia, Mahjoub Aouni, Hedi Harizi
ABSTRACT
The immune response is the first defense against pathogens; however, it is very sensitive and can be impacted on by agrochemicals such as carbamate and organophosphate pesticides widely present in the environment. To understand how pesticides can affect immune cell function in vitro, this study investigated the effects of chlorpyrifos (CPF) and carbendazim (CBZ), the most commonly used pesticides worldwide, on murine immune cell (i.e. macrophage) functions, including lysosomal enzyme activity and pro-inflammatory cytokines (IL-1β and TNFα) and nitric oxide (NO) production by isolated mouse peritoneal macrophages. This study showed for the first time that CPF and CBZ dose-relatedly reduced macrophage lysosomal enzyme activity and LPS-induced production of IL-1β, TNFα and NO. In general, the effects caused by CPF appeared more pronounced than those by CBZ. Collectively, these results demonstrated that CPF and CBZ exhibited marked immunomodulatory effects and could act as potent immunosuppressive factors in vitro. This inhibition of macrophage pro-inflammatory function may be an integral part of the underlying mode of action related to pesticide-induced immunosuppression.
Sigma-Aldrich
Lectin from Phaseolus vulgaris (red kidney bean), Phytohemagglutinin PHA-M, lyophilized powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.