MilliporeSigma
  • Investigations into the association between polymorphisms in the interleukin-10 gene and risk of early-onset preeclampsia.
Investigations into the association between polymorphisms in the interleukin-10 gene and risk of early-onset preeclampsia.
Genetics and molecular research : GMR (2015)
Liu QY, et al.