MilliporeSigma
  • Simple Preparation of Rhodococcus erythropolis DSM 44534 as Biocatalyst to Oxidize Diols into the Optically Active Lactones.
Simple Preparation of Rhodococcus erythropolis DSM 44534 as Biocatalyst to Oxidize Diols into the Optically Active Lactones.
Chirality (2016-08-09)
Enriqueta Martinez-Rojas, Teresa Olejniczak, Konrad Neumann, Leif-Alexander Garbe, Filip Boratyñski
ABSTRACT
In the current study, we present a green toolbox to produce ecological compounds like lactone moiety. Rhodococcus erythropolis DSM 44534 cells have been used to oxidize both decane-1,4-diol () and decane-1,5-diol () into the corresponding γ- () and δ-decalactones () with yield of 80% and enantiomeric excess (ee) = 75% and ee = 90%, respectively. Among oxidation of meso diols, (-)-(1S,5R)-cis-3-oxabicyclo[4.3.0]non-7-en-2-one (5a) with 56% yield and ee = 76% as well as (-)-(2R,3S)-cis-endo-3-oxabicyclo[2.2.1]dec-7-en-2-one (6a) with 100% yield and ee = 90% were formed. It is worth mentioning that R. erythropolis DSM 44534 grew in a mineral medium containing ethanol as the sole source of energy and carbon Chirality 28:623-627, 2016. © 2016 Wiley Periodicals, Inc.
Sigma-Aldrich
1,2-Cyclohexanedicarboxylic anhydride, predominantly cis, 95%